Learn More
MSC Kranzle Pressure Washer, 1,600 psi, 1.7 GPM, Electric, Cold Water
Catalog No. 502837864
Encompass
This cold water electric pressure washer is designed for industrial and commercial applications, suitable for professionals in fields such as hotel and building cleaning, auto detailing, HVAC maintenance, screen printing, and washing boats or vehicles.
  • Advanced pump design provides multi-hour bypass and run-dry capability, preventing accidental pump damage.
  • Dirt Killer turbo nozzle doubles cleaning efficiency.
  • Vario-jet nozzle combines the functions of four flat tip nozzles into one easy-to-use package.
  • Adjustable forged brass unloader.
  • 50-feet wire braided hose on a hose reel with safety couplers.
  • TEFC 1600 RPM electric 110V, 15A motor with GFI, can be plugged in anywhere.
Catalog No. 50-283-7864 Supplier MSC Supplier No. 36965143
This cold water electric pressure washer is designed for industrial and commercial applications, suitable for professionals in fields such as hotel and building cleaning, auto detailing, HVAC maintenance, screen printing, and washing boats or vehicles.
  • Advanced pump design provides multi-hour bypass and run-dry capability, preventing accidental pump damage.
  • Dirt Killer turbo nozzle doubles cleaning efficiency.
  • Vario-jet nozzle combines the functions of four flat tip nozzles into one easy-to-use package.
  • Adjustable forged brass unloader.
  • 50-feet wire braided hose on a hose reel with safety couplers.
  • TEFC 1600 RPM electric 110V, 15A motor with GFI, can be plugged in anywhere.