Learn More
Specifications
Specifications
| Certifications/Compliance | USP Class VI |
| For Use With (Application) | SIP Processing |
| Tubing Size | 3/4 x 1-1/2 in. |
| Hose Barb | 1/2 Hose Barb |
| Material (Seal) | Platinum-cured Silicone |
| Format | Free-floating |
| Product Type | ADCF SIP Connector |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.