Learn More
CPC™ Company MPC Body x MPC Back-to-Back Body Adapter
Catalog No. 05113243 Shop All Colder Products Company Products
Change view
Click to view available options
MPC body x MPC back-to-back body adapter
Catalog No.
05-113-243 MPC body x MPC back-to-back body adapter
1 options
Catalog No. 05-113-243 Supplier CPC™ Supplier No. MPC17C1739
Specifications
Color Amber
Material Polysulfone
Type Back-to-back Body Adapter
Pressure Up to 60 psi
Temperature (Metric) Max 149°C