Learn More
CPC™ HFC35 Polysulfone Hose Barb Coupling Inserts
Catalog No. 05113154 Shop All Colder Products Company Products
Change view
Click to view available options
Product Type:
Hose Barb Non-valved In-line Coupling Insert
Hose Barb Valved In-line Coupling Insert
Hose Barb:
1/2 in.
3/8 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Hose Barb
05-113-154 Hose Barb Valved In-line Coupling Insert 3/8 in.
05-113-155 Hose Barb Valved In-line Coupling Insert 1/2 in.
05-113-153 Hose Barb Non-valved In-line Coupling Insert 1/2 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 05-113-154 Supplier CPC™ Supplier No. HFCD22635
Free floating
  • Polysulfone couplings
  • Withstands demanding applications
  • UV resitant, able to withstand continued exposure to harmful UV rays
  • Ergonomic design
  • Operate in harsh environments in temperatures from -40°F to 280°F (-40°C to 138°C)
  • Pressure: Vacuum to 125psi (8.6 bar)
  • Large, shrouded thumb latch pad
  • Leads to high flow and exceptionally low spillage
Specifications
Material Polysulfone
Hose Barb 3/8 in.
Product Type Hose Barb Valved In-line Coupling Insert