Learn More
Bellco Glass, Inc. Coldfinger Assembly , 3L
SDP
Supplier:  Bellco Glass, Inc. 196402070
Encompass_Preferred
Used to generate a localized cold surface on the exterior of the finger. Commonly used for sublimation, and can also be used in re-flux or distillation operations. 3L. MADE IN THE USA
Catalog No. 50-194-4726