Learn More
MSC Perma-Patch Air Operated Diaphragm Pump, 55 Gallon Drum, 120 psi, 52 gpm
Catalog No. 502839638
Encompass
This air-operated diaphragm pump is engineered for efficient and precise application of sealants from 55-gallon drums. Designed for large-scale pavement projects, it streamlines the crack sealing and surface coating process, delivering consistent flow and professional results while minimizing manual labor.
  • High-efficiency pump kit with a precision wand for accurate application
  • Streamlines application, enabling continuous operation and reducing manual effort
  • Suitable for extensive road maintenance and diverse weather conditions
  • Compatible with asphalt repair products for crack sealing and surface coating
  • Features a maximum flow rate of 52 gpm and operating pressure of 120 psi
  • Constructed with an aluminum wetted housing and Hytrel diaphragm and valve materials
Catalog No. 50-283-9638 Supplier MSC Supplier No. 76052539
Add to Cart
Add to Cart
Request a Quote
This air-operated diaphragm pump is engineered for efficient and precise application of sealants from 55-gallon drums. Designed for large-scale pavement projects, it streamlines the crack sealing and surface coating process, delivering consistent flow and professional results while minimizing manual labor.
  • High-efficiency pump kit with a precision wand for accurate application
  • Streamlines application, enabling continuous operation and reducing manual effort
  • Suitable for extensive road maintenance and diverse weather conditions
  • Compatible with asphalt repair products for crack sealing and surface coating
  • Features a maximum flow rate of 52 gpm and operating pressure of 120 psi
  • Constructed with an aluminum wetted housing and Hytrel diaphragm and valve materials