Learn More
Shockwatch Div Mri ColdMark 10C/50F
SDP
Catalog No. NC2598325
Encompass_Preferred
ColdMark temperature indicator monitors temperatures during shipping and storage, indicating if goods experience unacceptable conditions. These reliable temperature indicators provide visual evidence of exposure to unacceptably low temperature levels, allowing you to determine if product quality has been compromised. Temperature threshold: 10C/50F. Accuracy +/-1C. Shelf life 2 years. Single-use.
Catalog No. NC2598325 Supplier Shockwatch Div Mri Supplier No. CM 10/50
ColdMark temperature indicator monitors temperatures during shipping and storage, indicating if goods experience unacceptable conditions. These reliable temperature indicators provide visual evidence of exposure to unacceptably low temperature levels, allowing you to determine if product quality has been compromised. Temperature threshold: 10C/50F. Accuracy +/-1C. Shelf life 2 years. Single-use.