Learn More
Mopec™ Coldplate Base
SDP
Catalog No. 22445436 Shop All Mopec Products
Change view
Click to view available options
Product Type:
Coldplate Base
Catalog No. Product Type
22-445-436 Coldplate Base
1 options
Catalog No. 22-445-436 Supplier Mopec™ Supplier No. BK600B
Add to Cart
Add to Cart
The Mopec™ Coldplate is designed to replace chilling sprays or ice blocks that are typically used to cool down tissue blocks before sectioning.
  • 36 tissue block capacity
  • Designed to take up minimal bench-top space
  • Removes the need for chilling sprays or ice blocks
  • Remains cold for up to 4 hours
  • No electrical components or compressors required
Specifications
Product Type Coldplate Base
For Use With (Equipment) Chilling Sprays, Ice Blocks