Learn More
Cole-Parmer Essentials Universal Pipette Tips with Filter, Sterile, 1000 μL; 10 Racks x 96 Tips
Catalog No. NC2015039 Shop All Cole-Parmer Products
Encompass_Preferred
Cole-Parmer sterile pipette tips are constructed of high-grade polypropylene and a polyethylene filter which prevents liquids and aerosols from entering the body of your pipette. Tips are 99% Bacterial Aerosol Filtration Efficient (BFE), ASTM F21012, ISO 10993-5 MRM, minimum essential media elution, and GLP compliant.
Catalog No. NC2015039 Supplier Cole-Parmer Supplier No. 7890138
Cole-Parmer sterile pipette tips are constructed of high-grade polypropylene and a polyethylene filter which prevents liquids and aerosols from entering the body of your pipette. Tips are 99% Bacterial Aerosol Filtration Efficient (BFE), ASTM F21012, ISO 10993-5 MRM, minimum essential media elution, and GLP compliant.