Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Polypropylene, Straight, Female Luer x 1/8-27 NPT; 25/PK
Catalog No. NC1285572
Encompass
These adapters transition between luer fittings and UNF or NPT threads. Both male and female luer connections are available. All threaded connections are female thread.
Catalog No. NC1285572 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3080000
These adapters transition between luer fittings and UNF or NPT threads. Both male and female luer connections are available. All threaded connections are female thread.