Learn More
Cole-Parmer Cole-Parmer Column Clamp, x-Large, Stainless Steel, 3.62" to 6.5" Grip Range
Catalog No. NC1315428 Shop All Cole-Parmer Products
Encompass_Preferred
Ideal for large cylindrical glassware and similar labware. The large, flat thumbscrew/worm-drive provides a firm hold. NOTE: An additional connector must be purchased in order to attach clamps to frames or other apparatus.
Catalog No. NC1315428 Supplier Cole-Parmer Supplier No. 0806610
Ideal for large cylindrical glassware and similar labware. The large, flat thumbscrew/worm-drive provides a firm hold. NOTE: An additional connector must be purchased in order to attach clamps to frames or other apparatus.