Learn More
Cole-Parmer Essentials Slot Sampler, 1 Opening, Sterile, HDPE, 970mm Length, 50mL; 20/PK
Catalog No. NC2228160 Shop All Cole-Parmer Products
Encompass_Preferred
These slot samplers provide highly representative sampling of powders, granules, and grains. They are suitable for sampling from all layers of the container, providing an accurate representation of the contents. The samplers are manufactured, assembled, and packed in a Class 100,000 Cleanroom to ensure cleanliness. Samplers are also sterilized via gamma irradiation (25-45 kGy) and individually bagged to reduce contamination.
This item is not returnable. View return policy
Catalog No. NC2228160 Supplier Cole-Parmer Supplier No. 0628622
These slot samplers provide highly representative sampling of powders, granules, and grains. They are suitable for sampling from all layers of the container, providing an accurate representation of the contents. The samplers are manufactured, assembled, and packed in a Class 100,000 Cleanroom to ensure cleanliness. Samplers are also sterilized via gamma irradiation (25-45 kGy) and individually bagged to reduce contamination.