Learn More
Cole-Parmer H2300-HC2 Mini CO2 Digital Incubator; 115 V
Catalog No. NC2004012 Shop All Cole-Parmer Products
Encompass_Preferred
The mini digital temperature and CO2 control incubators are only 13 inches wide, small enough to fit in most biological safety cabinets, conserving valuable bench space. A thick layer of high-efficiency insulation and a 12 VDC car adapter make this incubator ideal for transporting cells. The unit can maintain set temperature levels within 10% for up to one hour after power has been disconnected. Intuitive control panel and large LED displays allow you to digitally adjust both temperature and CO² percentages and view in real time. Another function on the panel provides simple in-lab calibration of temperature or CO². You can add up to 80% humidity to the chamber by inserting a stainless steel reservoir.
Catalog No. NC2004012 Supplier Cole-Parmer Supplier No. 0360805
The mini digital temperature and CO2 control incubators are only 13 inches wide, small enough to fit in most biological safety cabinets, conserving valuable bench space. A thick layer of high-efficiency insulation and a 12 VDC car adapter make this incubator ideal for transporting cells. The unit can maintain set temperature levels within 10% for up to one hour after power has been disconnected. Intuitive control panel and large LED displays allow you to digitally adjust both temperature and CO² percentages and view in real time. Another function on the panel provides simple in-lab calibration of temperature or CO². You can add up to 80% humidity to the chamber by inserting a stainless steel reservoir.