Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Polypropylene, Straight, Sanitary Ladish Clamp to Threaded Adapter, 1"" x 3/8"" NPT(F)
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3180602/DEL
Encompass
The sanitary clamp fittings may be autoclaved. The PVDF material tolerates higher temperatures better than polypropylene and performs more consistently after repeated autoclaving.
Catalog No. NC1374370