Learn More
Cole-Parmer Cole-Parmer Stainless Steel Tweezers, Reverse Action, Fiber Grip, Straight Bent Tips, 165mm
Catalog No. NC3731871 Shop All Cole-Parmer Products
Encompass_Preferred
Corrosion resistantExcellent tip hardnessMagnetic stainless steel constructionSelf-lock mechanism is ideal for solderingBent tips helps enhance visibilityProvides access to small components in confined areasHeat-resistant fiber grips protect your hands from heat
Catalog No. NC3731871 Supplier Cole-Parmer Supplier No. 0739816
Corrosion resistantExcellent tip hardnessMagnetic stainless steel constructionSelf-lock mechanism is ideal for solderingBent tips helps enhance visibilityProvides access to small components in confined areasHeat-resistant fiber grips protect your hands from heat