Learn More
Cole-Parmer Cole-Parmer Two-Position Chilling/Heating Block, 100 VAC
Supplier:  Cole-Parmer 4417506
Encompass_Preferred
Replace your ice buckets or inconvenient freezer blocks with this compact, economical benchtop chilling/heating block. Features include a two-line LCD that reads actual and set point temperatures, 30-day countdown timer with alarm and auto/off, built-in data logger, and RS-232 I/O port for bidirectional communication with an IBM compatible computer. Energy-efficient Peltier design chills or heats biological samples from -10 to 100C with ±1C digital control.
Catalog No. NC1107062