Learn More
Cole-Parmer Cole-Parmer UN/DOT Liquid Waste Container, S70 cap; 20 L; 1/ea
Catalog No. NC1589973 Shop All Cole-Parmer Products
Encompass_Preferred
Sturdy, rectangular UN/DOT carboys are easy to fill and ship for disposal. Safely use these single-use carboys to transfer and store liquid waste, use as waste traps with automated instruments, or as part of your HPLC system. UN rating: 3H1/Y1.8/100
Catalog No. NC1589973 Supplier Cole-Parmer Supplier No. 0605999
Sturdy, rectangular UN/DOT carboys are easy to fill and ship for disposal. Safely use these single-use carboys to transfer and store liquid waste, use as waste traps with automated instruments, or as part of your HPLC system. UN rating: 3H1/Y1.8/100