Learn More
Cole-Parmer Cole-Parmer Watch Glass 100mm 12/pk
Supplier:  Cole-Parmer 3450858
Encompass_Preferred
Manufactured from clear glass sheet (1.8 to 2.0mm thick), these watch glasses are fire ground and have polished edges. Concave shape prevents sliding and return of condensation to the beaker.
Catalog No. NC1344483