Learn More
Aviva Systems Biology Corp COLEC12 Antibody - middle region (ARP50285_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50285_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against COLEC12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human COLEC12. Alternate names: CLP1, NSR2, SCARA4, SRCLApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60308