Learn More
R3 Safety™ Coleman Flotation Vests
Catalog No. 19819921 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Buoyancy:
15.5 lb.
17.5 lb.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Buoyancy
19-819-921 15.5 lb.
19-819-916 17.5 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-819-921 Supplier R3 Safety™ Supplier No. 14960
This item is not returnable. View return policy
Specifications
Product Type Flotation Vest
Material Nylon
Closure Type Hook and loop
Buoyancy 15.5 lb.
No. of Pockets 4.0
Certifications/Compliance USCG Type I vest
For Use With (Application) Designed with adjustable side tabs that slide quickly for a customizable, contoured fit