Learn More
Medchemexpress LLC Colesevelam hydrochl 25mg | 182815-44-7 | 25MG
SDP
Catalog No. 5000240990
Encompass_Preferred
Colesevelam (hydrochloride) is a bile acid sequestrant that bind bile acids in the gut Colesevelam (hydrochloride) inhibits bile acid reabsorption leading to increased bile acid synthesis and reduced cholesterol levels in patients with cholestatic pruritus and Crohn s disease
Catalog No. 50-002-40990 Supplier Medchemexpress LLC Supplier No. HY10653925MG
Colesevelam (hydrochloride) is a bile acid sequestrant that bind bile acids in the gut Colesevelam (hydrochloride) inhibits bile acid reabsorption leading to increased bile acid synthesis and reduced cholesterol levels in patients with cholestatic pruritus and Crohn s disease