Learn More
Medchemexpress LLC Colesevelam (hydrochloride) | 182815-44-7 | 95.0% | 3694.64 | (C3H8NCl)2.(C9H20N2OCl2).(C13H28NCl)7.(C12H28N2Cl2)6 | 100 MG
SDP
Catalog No. 5000242090
Encompass_Preferred
Colesevelam hydrochloride is a nonabsorbed bile acid sequestrant used in research to study bile acid binding and cholesterol-lowering mechanisms. It binds bile acids in the gut and inhibits bile acid reabsorption, which can increase bile acid synthesis and reduce serum cholesterol. Supplied as a solid for laboratory use with supporting COA and SDS documents.
Catalog No. 50-002-42090 Supplier Medchemexpress LLC Supplier No. HY106539100MG
Colesevelam hydrochloride is a nonabsorbed bile acid sequestrant used in research to study bile acid binding and cholesterol-lowering mechanisms. It binds bile acids in the gut and inhibits bile acid reabsorption, which can increase bile acid synthesis and reduce serum cholesterol. Supplied as a solid for laboratory use with supporting COA and SDS documents.