Learn More
Medchemexpress LLC Colfosceril miristate-d27 (DMPC-d27) | 185906-03-0 | 99.0% | 705.10 | 1 MG
SDP
Catalog No. 5000363004
Encompass_Preferred
1-Myristoyl-2-myristoyl-sn-glycero-3-phosphocholine-d27 is deuterium labeled 1-Myristoyl-2-myristoyl-sn-glycero-3-phosphocholine. It is sold for research purposes only and not for patient use. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-63004 Supplier Medchemexpress LLC Supplier No. HY146881S1MG
Add to Cart
Add to Cart
1-Myristoyl-2-myristoyl-sn-glycero-3-phosphocholine-d27 is deuterium labeled 1-Myristoyl-2-myristoyl-sn-glycero-3-phosphocholine. It is sold for research purposes only and not for patient use. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.