Learn More
Enterprise Technology Solutions Colgate-Palmolive Company Cleaner, Multipurpose, 1 Gallon, Lavender
SDP
Catalog No. 5000830927
Encompass_Preferred
Fabuloso Professional is an all-purpose cleaner and degreaser that is effective for a wide variety of cleaning applications. This formula deep-cleans to remove grease and soils while leaving a long-lasting fragrance. Its neutral pH makes the cleaner safe for surfaces. Rinsing step is not required in floor cleaning. To clean floors, kitchens, bathrooms and walls, mix 2 oz. of Fabuloso with 1 gallon of water to remove grease, grime and dirt. Use in toilet bowls to clean and deodorize. Cleaner includes bilingual labeling.
Catalog No. 50-008-30927 Supplier Enterprise Technology Solutions Supplier No. CPCUS05253A
Fabuloso Professional is an all-purpose cleaner and degreaser that is effective for a wide variety of cleaning applications. This formula deep-cleans to remove grease and soils while leaving a long-lasting fragrance. Its neutral pH makes the cleaner safe for surfaces. Rinsing step is not required in floor cleaning. To clean floors, kitchens, bathrooms and walls, mix 2 oz. of Fabuloso with 1 gallon of water to remove grease, grime and dirt. Use in toilet bowls to clean and deodorize. Cleaner includes bilingual labeling.