Learn More
Fine Science Tools Colibri Retractors, Stainless Steel, 4 cm
SDP
Catalog No. NC9128321
Encompass
Product: Colibri Retractor - Wire/4cm/1.5cm Maximum Spread/ Teeth: Wire/ Teeth Length: 4mm/ Teeth Width: 14mm/ Maximum Spread: 1.5cm/ Alloy: Stainless Steel/ Length: 4cm/ Feature: Slightly Curved/ Unit: Each
Catalog No. NC9128321 Supplier Fine Science Tools Supplier No. 1700004
Product: Colibri Retractor - Wire/4cm/1.5cm Maximum Spread/ Teeth: Wire/ Teeth Length: 4mm/ Teeth Width: 14mm/ Maximum Spread: 1.5cm/ Alloy: Stainless Steel/ Length: 4cm/ Feature: Slightly Curved/ Unit: Each