Learn More
Fine Science Tools Colibri Retractors, Stainless Steel, 3 cm
SDP
Catalog No. NC0518945
Encompass
Product: Colibri Retractor - Wire/3cm/1.2cm Maximum Spread/ Teeth: Wire/ Teeth Length: 3mm/ Teeth Width: 4mm/ Maximum Spread: 1.2cm/ Alloy: Stainless Steel/ Length: 3cm/ Feature: Slightly Curved/ Unit: Each
Catalog No. NC0518945 Supplier Fine Science Tools Supplier No. 1700002
Product: Colibri Retractor - Wire/3cm/1.2cm Maximum Spread/ Teeth: Wire/ Teeth Length: 3mm/ Teeth Width: 4mm/ Maximum Spread: 1.2cm/ Alloy: Stainless Steel/ Length: 3cm/ Feature: Slightly Curved/ Unit: Each