Learn More
World Precision Instrument Colibri Tying Forceps, 11.5cm, Straight, Titanium
SDP
Catalog No. 502538304
Encompass
WPI Titanium Forceps with Tying Platform are straight forceps 11.5 cm (4.5") long, and they come with 4 mm, 5 mm or 6 mm tying platforms. Forceps with a tying platform are designed to grasp, and manipulate tissues or objects during surgical procedures. They flat tying platform is near the tips where suture can be tied. This gives you a stable area for suturing or ligating. The slender handles are textured for a firm grip, and the hole patterns in the handle makes the instrument incredibly lightweight. All three sizes are made of titanium, which is 40% lighter than stainless steel.  Titanium is 100% non-corrosive, great for researchers doing seawater procedures. Titanium is 100% non-magnetic, making it MRI compatible.
Catalog No. 50-253-8304 Supplier World Precision Instrument Supplier No. WP500097
WPI Titanium Forceps with Tying Platform are straight forceps 11.5 cm (4.5") long, and they come with 4 mm, 5 mm or 6 mm tying platforms. Forceps with a tying platform are designed to grasp, and manipulate tissues or objects during surgical procedures. They flat tying platform is near the tips where suture can be tied. This gives you a stable area for suturing or ligating. The slender handles are textured for a firm grip, and the hole patterns in the handle makes the instrument incredibly lightweight. All three sizes are made of titanium, which is 40% lighter than stainless steel.  Titanium is 100% non-corrosive, great for researchers doing seawater procedures. Titanium is 100% non-magnetic, making it MRI compatible.