Learn More
Lovibond™ Coliform / E.coli Test Kit
Catalog No. 09053296 Shop All Tintometer Group Products
Change view
Click to view available options
Volume (Metric):
100 mL
Catalog No. Volume (Metric)
09-053-296 100 mL
1 options
Catalog No. 09-053-296 Supplier Lovibond™ Supplier No. 56K009701
Add to Cart
Add to Cart
Comes complete with everything needed to measure coliforms and E.Coli in water samples in a single step.
  • Early identification of bacterial contamination
  • Simple one step procedure
  • Coliforms and E.coli in one test
  • 100 mL sample (regulatory reporting)
  • In the case of coliforms, the solution turns green-blue. E. coli bacteria are detected by fluorescence under UV light.
Specifications
pH Range 0 to 14
Volume (Metric) 100 mL
Type Coliform Test