Learn More
Thermco Products Inc NIST compliant, factory certified glass Spirit thermometer, 34.5 deg./46 deg.C, 0.1C div, 76mm imm, certified at 37 deg.C, 44.5 deg.C, 240mm
Catalog No. NC0633533
Encompass_Preferred
These spirit-filled glass thermometers meet EPA requirements. Coliform incubator thermometers are designed to use in incubators or baths during coliform tests. Use them to monitor equipment. Each includes an individual serial number and certificate of accuracy. The ACC713BLS is certified and traceable to N.I.S.T.
Catalog No. NC0633533 Supplier Thermco Products Inc Supplier No. ACC713BLSFC
These spirit-filled glass thermometers meet EPA requirements. Coliform incubator thermometers are designed to use in incubators or baths during coliform tests. Use them to monitor equipment. Each includes an individual serial number and certificate of accuracy. The ACC713BLS is certified and traceable to N.I.S.T.