Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aldon Corporation Coliform Test Powder Kit
GREENER_CHOICESCI_ED

Catalog No. S06786 Shop All Aldon Products
Change view
Click to view available options
Class Size:
1 Lab Group
5 Lab Groups
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S06786 5 Lab Groups
S06785 1 Lab Group
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S06786 Supplier Aldon Corporation Supplier No. IS5056
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Coliform test powder kit is an extremely easy way to test your water sample for coliforms.

  • Simply add a small amount of collected water to the sample tube and incubate for 24 to 48 hours.
  • A distinct color change from purple to yellow indicates that coliforms are present.

Specifications

Grade 9 to 12
Includes Instructions and 5 Coliform Powder Test Kits
Class Size 5 Lab Groups
Product Type Coliform Powder Test Tube
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.