Learn More
Hach Company MPN Tube Rack
Catalog No. 501994725 Shop All Hach Company Products
Encompass
Powder coated steel rack holds 25 Hach microbiology MPN tubes (19 mm outside diameter). Holes keyed with letters, numbers for identification. Withstands up to 475°F.
Catalog No. 50-199-4725 Supplier Hach Company Supplier No. 221500
Powder coated steel rack holds 25 Hach microbiology MPN tubes (19 mm outside diameter). Holes keyed with letters, numbers for identification. Withstands up to 475°F.