Learn More
Description
- Each cartridge contains 50 discs that are spring-loaded for smooth and reliable dispensing
- Easy-to-use testing discs
Specifications
Specifications
| Concentration | 10 mcg |
| Product Type | Antimicrobial Susceptibility Disc |
| Target Organism Class | Multidrug-resistant, Gram-negative Organisms |
| Test Type | Antimicrobial Susceptibility |
| Packaging Type | Cartridge |
| Quantity | 50 Discs |
| Species | Carbapenem-Resistant Enterobacteriaceae (CRE) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
