Learn More
Apexbio Technology LLC Colistin Methanesulfonate (sodium salt)(Synonyms: Colistimethate sodium, Colistin sodium methanesulfonate, CMS, Sodium colistimethate), 5g, CAS: 8068-28-8.
SDP
Catalog No. 5000060521
Encompass_Preferred
Colistin methanesulfonate (sodium salt CAS 8068-28-8) is an inactive prodrug of colistin (polymyxin E) a polypeptide antibiotic derived from Paenibacillus polymyxa strains It targets Gram-negative bacteria by disrupting the integrity of bacterial outer membranes leading to bacterial cell death In vitro analyses indicate concentration-dependent bactericidal activity against Pseudomonas aeruginosa requiring significantly elevated levels above minimum inhibitory concentrations (MIC) for complete microbial eradication It serves primarily as a tool compound for exploring antimicrobial mechanisms and resistance dynamics in Gram-negative pathogens
Catalog No. 50-000-60521 Supplier Apexbio Technology LLC Supplier No. C30465000
Colistin methanesulfonate (sodium salt CAS 8068-28-8) is an inactive prodrug of colistin (polymyxin E) a polypeptide antibiotic derived from Paenibacillus polymyxa strains It targets Gram-negative bacteria by disrupting the integrity of bacterial outer membranes leading to bacterial cell death In vitro analyses indicate concentration-dependent bactericidal activity against Pseudomonas aeruginosa requiring significantly elevated levels above minimum inhibitory concentrations (MIC) for complete microbial eradication It serves primarily as a tool compound for exploring antimicrobial mechanisms and resistance dynamics in Gram-negative pathogens