Learn More
Medchemexpress LLC Colistin (sulfate) | 1264-72-8 | ≥98.0% | C52H98N16O13.2.5H2O4S | 100 MG
SDP
Catalog No. 5000433479
Encompass_Preferred
Colistin sulfate is a polypeptide antibiotic effective against gram-negative bacteria. It functions by binding to lipopolysaccharides and phospholipids in the outer cell membrane, competitively displacing divalent cations. This leads to a detergent-like mechanism that is bactericidal to gram-negative bacteria. It is characterized by rapid bacterial killing, a narrow spectrum of activity, and slow development of resistance.
  • Inhibits gram-negative bacteria
  • Binds to lipopolysaccharides and phospholipids
  • Displaces divalent cations (calcium and magnesium)
  • Bactericidal through a detergent-like mechanism
  • Rapid bacterial killing
  • Narrow spectrum of activity
  • Slow development of resistance
Catalog No. 50-004-33479 Supplier Medchemexpress LLC Supplier No. HYA0089100MG
Colistin sulfate is a polypeptide antibiotic effective against gram-negative bacteria. It functions by binding to lipopolysaccharides and phospholipids in the outer cell membrane, competitively displacing divalent cations. This leads to a detergent-like mechanism that is bactericidal to gram-negative bacteria. It is characterized by rapid bacterial killing, a narrow spectrum of activity, and slow development of resistance.
  • Inhibits gram-negative bacteria
  • Binds to lipopolysaccharides and phospholipids
  • Displaces divalent cations (calcium and magnesium)
  • Bactericidal through a detergent-like mechanism
  • Rapid bacterial killing
  • Narrow spectrum of activity
  • Slow development of resistance