Learn More
Medchemexpress LLC Colistin sulfate | 1264-72-8 | MFCD00216503 | >98.0% | C52H98N16O15·2.5H2SO4 | 5 G
SDP
Catalog No. 5000433557
Encompass_Preferred
Colistin sulfate is a polypeptide antibiotic which inhibits gram-negative bacteria by binding to lipopolysaccharides and phospholipids in the outer cell membrane of gram-negative bacteria.
  • Polypeptide antibiotic
  • Inhibits gram-negative bacteria
  • Purity of >98%
  • Powder storage at -20°C for 3 years or 4°C for 2 years
  • In solvent storage at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-004-33557 Supplier Medchemexpress LLC Supplier No. HYA00895G
Colistin sulfate is a polypeptide antibiotic which inhibits gram-negative bacteria by binding to lipopolysaccharides and phospholipids in the outer cell membrane of gram-negative bacteria.
  • Polypeptide antibiotic
  • Inhibits gram-negative bacteria
  • Purity of >98%
  • Powder storage at -20°C for 3 years or 4°C for 2 years
  • In solvent storage at -80°C for 6 months or -20°C for 1 month