Learn More
Selleck Chemical LLC Colistin Sulfate S4029-50mg
SDP
Catalog No. 503109917
Encompass_Preferred
Colistin (Polymixin E) is a cyclic cationic decapeptide linked to a fatty acid side chain it belongs to a group of similarly structured bacterial antimicrobial peptides
Catalog No. 50-310-9917 Supplier Selleck Chemical LLC Supplier No. S402950MG
Colistin (Polymixin E) is a cyclic cationic decapeptide linked to a fatty acid side chain it belongs to a group of similarly structured bacterial antimicrobial peptides