Learn More
Selleck Chemical LLC Colistin Sulfate S4029-1g
SDP
Catalog No. 503111232
Encompass_Preferred
Colistin (Polymixin E) is a cyclic cationic decapeptide linked to a fatty acid side chain it belongs to a group of similarly structured bacterial antimicrobial peptides
Catalog No. 50-311-1232 Supplier Selleck Chemical LLC Supplier No. S40291G
Colistin (Polymixin E) is a cyclic cationic decapeptide linked to a fatty acid side chain it belongs to a group of similarly structured bacterial antimicrobial peptides