Learn More
Medchemexpress LLC HY-A0089 500mg Medchemexpress, Colistin (sulfate) Polymyxin E Sulfate CAS:1264-72-8 Purity:98%
SDP
Catalog No. 501639112
Encompass_Preferred
Medchemexpress,HY-A0089 500mg Colistin (sulfate) Polymyxin E Sulfate CAS:1264-72-8 Formula:C52H100N16O17S Colistin, a polypeptide antibiotic, is bactericidal to gram-negative bacteria by interfering with the structure and function of the outer and cytoplasmic membranes of bacteria. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-9112 Supplier Medchemexpress LLC Supplier No. HYA0089500MG
Medchemexpress,HY-A0089 500mg Colistin (sulfate) Polymyxin E Sulfate CAS:1264-72-8 Formula:C52H100N16O17S Colistin, a polypeptide antibiotic, is bactericidal to gram-negative bacteria by interfering with the structure and function of the outer and cytoplasmic membranes of bacteria. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.