Learn More
Enzo Life Sciences Colistin . sulfate (500mg). CAS: 1264-72-8
SDP
Supplier:  Enzo Life Sciences ALX380272M500
Encompass_Preferred
Polymyxin antibiotic. Binds to lipids on the cell cytoplasmic membrane. Disrupts cell wall integrity. Effective against Gram-negative bacteria. Alternative name: Polymyxin E . sulfate. Solubility: Soluble in water (50mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-9005