Learn More
Abcam Colistin sulfate, Cationic polypeptide antibiotic, 100MG
Catalog No. 502706119
Encompass_Preferred
MW 1155.4 Da. Cationic polypeptide antibiotic. Interacts with bacterial cell membranes displacing counter ions in lipopolysaccharides solubilizing the membrane. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6119 Supplier Abcam Supplier No. AB143385100MG
MW 1155.4 Da. Cationic polypeptide antibiotic. Interacts with bacterial cell membranes displacing counter ions in lipopolysaccharides solubilizing the membrane. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement