Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Colistin sulfate salt

Catalog No. AC455392500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC455392500 250 mg
AC455390010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC455392500 Supplier Thermo Scientific Chemicals Supplier No. 455392500
  • $120.00
    $96.65 / Each of 1

    Save $23.35

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Colistin Sulfate salt (2:5), CAS # 1264-72-8, (synonym: Polymyxin E Complex), is a complex mixture of antibiotics, with Colistin A and B being the primary components. It has anti-bacterial activity against Gram-positive and Gram-negative bacteria and may also be used as a neuromuscular blocker.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description
• Colistin Sulfate salt is a stable, cationic, water-soluble, white powder composed of over 30 components
• Originally isolated from B. polymyxa, it inhibits the growth of the Gram-negative (e.g. E. coli, P. aeruginosa, P. fluorescens, and S. enterica), and Gram-positive bacteria (e.g. L. lactis, P. polymyxa, P. acidilactici, and S. aureus) by permeabilizing the bacterial cell membrane

Applications
• Colistin Sulfate has potent anti-endotoxin activity rendered through its ability to bind lippopolysaccharides that make up bacterial endotoxins
• This compound is often used in the management of multi-drug resistant Gram-negative infections

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1264-72-8

Specifications

Type Colistin Sulfate salt
Quantity 250 mg
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in methanol, insoluble in acetone, diethyl ether
Percent Purity ≥77% Sum of Polymyxins (HPLC)
Chemical Name or Material Colistin sulfate salt
Hazard Category Danger
Hazard Statement GHS H Statement Toxic if swallowed.
Precautionary Statement GHS P Statement Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.