Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Colistin Sulfate Salt, MP Biomedicals

Catalog No. ICN19415725 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19415725 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19415725 Supplier MP Biomedicals, Inc Supplier No. 0219415725
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Polypeptide antibiotic that binds to and disrupts the cytoplasmic membrane of Gram-negative bacteria.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1264-72-8

Specifications

Physical Form Powder
Quantity 25 mg
Grade Not Available
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.