Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Colistin, sulfate salt, MP Biomedicals™

Catalog No. ICN19415783 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
0.25 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19415783 0.25 g
ICN19415791 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19415783 Supplier MP Biomedicals, Inc Supplier No. 0219415783
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1264-72-8

Specifications

Color Brown
Quantity 0.25 g
Physical Form Powder
Chemical Name or Material Colistin, sulfate salt
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.