Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Colistin Sulfate, USP Grade, G-Biosciences
SCI_ED

Catalog No. S979073
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S979073 25 g
S666999 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S979073 Supplier G-Biosciences Supplier No. RC987
Add to Cart
Add to Cart

Colistin is a cyclic polypeptide antibiotic of the polymyxin family. It targets the cell membrane of bacteria, which ultimately leads to an altered cell permeability and cell death. It primarily targets gram-negative bacteria.

Colistin has been used to study mannose-resistant haemagglutination, antibiotic resistance, and nephrotoxicity.

Chemical Identifiers

Molecular Weight (g/mol) 2801.26 g/mol

Specifications

Boiling Point 1536.8°C
Quantity 25 g
Percent Purity >98%
Grade USP
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.