Learn More
IDEXX REFERENCE LABORATORIES Colisure Test, Total Coliforms and E. coli, 200 tests per pack, incubation time 24 hours, read window up to 48 hours, unit-dosed packaging
Catalog No. NC2062137
Encompass
This supplier will only sell to Pharmaceutical and Manufacturing companies. They will not ship to Universities or Hospitals.
Colisure Test, Total Coliforms and E. coli, 200 tests per pack, incubation time 24 hours, read window up to 48 hours, unit-dosed packaging
Catalog No. NC2062137 Supplier IDEXX REFERENCE LABORATORIES Supplier No. 981317400
This supplier will only sell to Pharmaceutical and Manufacturing companies. They will not ship to Universities or Hospitals.
Colisure Test, Total Coliforms and E. coli, 200 tests per pack, incubation time 24 hours, read window up to 48 hours, unit-dosed packaging