Learn More
NEOGEN Colitag™ P/A Water Test Kit, Pack of 20
SDP
Supplier:  NEOGEN 700002856
Encompass_Preferred
Colitag™ uses a U.S. EPA-approved selective and differential medium to detect total coliforms and E. coli in water samples within 16–48 hours. Designed for use by personnel familiar with the appropriate aseptic techniques, NEOGEN recommends and offers training for users without advanced knowledge of microbiology.Approvals: EPA: Federal Register/vol. 74, No. 216/Tuesday, November 10, 2009/Rules and RegulationsModified Colitag™ is US EPA approved for compliance monitoring of public water systems as required by the Total Coliform Rule.Limit of Detection: 1 cfu in a 100 mL sampleFor more information visit https://www.neogen.com/solutions/Microbiology/colitag
Catalog No. 50-201-5139