Learn More
Medchemexpress LLC Colivelin TFA | 2803948-60-7 | 99.5% | 2759.12 | 25 MG
SDP
Catalog No. 5000426825
Encompass_Preferred
Colivelin TFA is a brain-penetrant neuroprotective peptide that acts as a potent activator of STAT3, suppressing neuronal death in vitro. It demonstrates long-term beneficial effects against neurotoxicity, amyloid-beta (Aβ) deposition, neuronal apoptosis, and deficits in synaptic plasticity associated with neurodegenerative diseases. This compound holds potential for the treatment of Alzheimer's disease and ischemic brain injury.
  • Neuroprotective properties
  • Potent activator of STAT3
  • Suppresses neuronal death
  • Exhibits long-term beneficial effects against neurotoxicity
  • Reduces amyloid-beta (Aβ) deposition
  • Prevents neuronal apoptosis
  • Addresses synaptic plasticity deficits
  • Potential treatment for Alzheimer's disease
  • Potential treatment for ischemic brain injury
Catalog No. 50-004-26825 Supplier Medchemexpress LLC Supplier No. 25MGHYP1061A
Colivelin TFA is a brain-penetrant neuroprotective peptide that acts as a potent activator of STAT3, suppressing neuronal death in vitro. It demonstrates long-term beneficial effects against neurotoxicity, amyloid-beta (Aβ) deposition, neuronal apoptosis, and deficits in synaptic plasticity associated with neurodegenerative diseases. This compound holds potential for the treatment of Alzheimer's disease and ischemic brain injury.
  • Neuroprotective properties
  • Potent activator of STAT3
  • Suppresses neuronal death
  • Exhibits long-term beneficial effects against neurotoxicity
  • Reduces amyloid-beta (Aβ) deposition
  • Prevents neuronal apoptosis
  • Addresses synaptic plasticity deficits
  • Potential treatment for Alzheimer's disease
  • Potential treatment for ischemic brain injury