Learn More
Medchemexpress LLC Colivelin TFA | 2803948-60-7 | 99.8% | 2759.12 | 5 MG
SDP
Catalog No. 5000425633
Encompass_Preferred
Colivelin TFA is a brain-penetrant neuroprotective peptide and a potent activator of STAT3. It suppresses neuronal death in vitro and offers long-term benefits against neurotoxicity, Aβ deposition, neuronal apoptosis, and synaptic plasticity deficits in neurodegenerative disease. It has potential for treating Alzheimer's disease and ischemic brain injury.
  • Suppresses neuronal death and activates neuroprotection through Ca2+/calmodulin-dependent protein kinase IV and STAT3 pathways.
  • Reverses caspase3, Bax, and Bcl-2 expressions.
  • Increases p-STAT3 protein levels.
  • Suppresses spatial working memory impairment and antagonizes hippocampal neuronal loss.
  • Improves motor/cognitive function and reduces lesion volume/neurological deficits post-MCAO.
  • Protects against cholinotoxin-induced amnesia and ischemic brain injury.
Catalog No. 50-004-25633 Supplier Medchemexpress LLC Supplier No. HYP1061A5MG
Colivelin TFA is a brain-penetrant neuroprotective peptide and a potent activator of STAT3. It suppresses neuronal death in vitro and offers long-term benefits against neurotoxicity, Aβ deposition, neuronal apoptosis, and synaptic plasticity deficits in neurodegenerative disease. It has potential for treating Alzheimer's disease and ischemic brain injury.
  • Suppresses neuronal death and activates neuroprotection through Ca2+/calmodulin-dependent protein kinase IV and STAT3 pathways.
  • Reverses caspase3, Bax, and Bcl-2 expressions.
  • Increases p-STAT3 protein levels.
  • Suppresses spatial working memory impairment and antagonizes hippocampal neuronal loss.
  • Improves motor/cognitive function and reduces lesion volume/neurological deficits post-MCAO.
  • Protects against cholinotoxin-induced amnesia and ischemic brain injury.