Learn More
Ibidi USA Collagen 1: #1.5 polymer coverslip, sterilized
SDP
Catalog No. NC2018031
Encompass
A 6 channel µ-Slide suitable for flow experiments and for immunofluorescence assays. All-in-one chamber that simplifies immunofluorescence protocols. Homogeneous cell distribution over the channel surface, regardless of handling practices. Cost-effective expirements with small numbers of cells and low volumes of reagents. Pcs./Box: 15 (individually packed)Surface Modification: Collagen I: #1.5 polymer coverslip, sterilized
This item is not returnable. View return policy
Catalog No. NC2018031 Supplier Ibidi USA Supplier No. 80609
A 6 channel µ-Slide suitable for flow experiments and for immunofluorescence assays. All-in-one chamber that simplifies immunofluorescence protocols. Homogeneous cell distribution over the channel surface, regardless of handling practices. Cost-effective expirements with small numbers of cells and low volumes of reagents. Pcs./Box: 15 (individually packed)Surface Modification: Collagen I: #1.5 polymer coverslip, sterilized