Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Collagen alpha-3(IV) chain (tumstatin), rat, N-terminal human Fc fusion | >90.0% | 50 UG
SDP

Catalog No. 5000292541
Encompass_Preferred

Recombinant rat Collagen alpha-3(IV) chain fragment (tumstatin) expressed in HEK293 cells and fused to an N-terminal human Fc tag. Supplied as a filtered PBS solution, this partial construct (G1426-H1670) is suitable for ELISA, antibody generation, and functional assays.

  • Expressed in HEK293 cells with N-terminal human Fc tag.
  • Partial COL4A3 fragment corresponding to residues G1426-H1670.
  • Molecular weight approximately 59 kDa.
  • Purity >90% by reducing SDS-PAGE.
  • Supplied as 0.22 μm filtered PBS solution, pH 7.4.
  • Endotoxin <1 EU/μg (LAL method).
  • Store at -80°C; shipped on dry ice.

Catalog No. 50-002-92541 Supplier Medchemexpress LLC Supplier No. HYP7568350UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant rat Collagen alpha-3(IV) chain fragment (tumstatin) expressed in HEK293 cells and fused to an N-terminal human Fc tag. Supplied as a filtered PBS solution, this partial construct (G1426-H1670) is suitable for ELISA, antibody generation, and functional assays.

  • Expressed in HEK293 cells with N-terminal human Fc tag.
  • Partial COL4A3 fragment corresponding to residues G1426-H1670.
  • Molecular weight approximately 59 kDa.
  • Purity >90% by reducing SDS-PAGE.
  • Supplied as 0.22 μm filtered PBS solution, pH 7.4.
  • Endotoxin <1 EU/μg (LAL method).
  • Store at -80°C; shipped on dry ice.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.